Assay Details
Target Gene Details
Entrez Gene ID: | 11328 |
Gene Name: | FK506 binding protein 9 |
Gene Aliases: |
FKBP60, FKBP63, PPIase |
Location: |
Chr.7:32957404-33006931 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP9 | NM_001284341.1 | 1 | 100 | NP_001271270.1 |
| NM_007270.4 | 1 | 100 | NP_009201.2 | |
| XM_011515115.2 | 1 | 100 | XP_011513417.1 | |
| AK075331.1 | 1 | 64 | ||
| AK128597.1 | ||||
| AK293954.1 | 1 | 30 | ||
| AK294266.1 | 1 | 30 | ||
| AK300328.1 | 1 | 63 | ||
| BC101723.1 | 1 | 28 | AAI01724.1 | |
| BC112053.1 | 1 | 28 | AAI12054.1 | |
| DA234297.1 | 1 | 100 | ||
| DC317549.1 | 1 | 137 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion |
|
| nsv1029220 | Chr.7:32881872 - 33222578 on Build GRCh38 | Gain |
|
| esv2734229 | Chr.7:32916268 - 33093711 on Build GRCh38 | Deletion |
|
| nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion |
|
| nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion |
|
| nsv1033096 | Chr.7:32925428 - 32999594 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs527879179] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map