Assay Details
Target Gene Details
Entrez Gene ID: | 221806 |
Gene Name: | von Willebrand factor D and EGF domains |
Gene Aliases: |
- |
Location: |
Chr.7:12330883-12404226 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWDE | NM_001135924.1 | NP_001129396.1 | ||
| XM_005249641.1 | XP_005249698.1 | |||
| XM_011515174.1 | XP_011513476.1 | |||
| XM_011515175.1 | XP_011513477.1 | |||
| XM_017011828.1 | XP_016867317.1 | |||
| XM_017011829.1 | XP_016867318.1 | |||
| AK027618.1 | BAB55237.1 | |||
| BC144319.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6272n100 | Chr.7:12311969 - 12404864 on Build GRCh38 | Loss |
|
| nsv1019021 | Chr.7:12270819 - 12335890 on Build GRCh38 | Loss |
|
| esv3612249 | Chr.7:12332122 - 12356965 on Build GRCh38 | Loss |
|
| nsv464371 | Chr.7:12332175 - 12395145 on Build GRCh38 | Gain |
|
| nsv606199 | Chr.7:11466516 - 12447033 on Build GRCh38 | Gain |
|

Genomic Map