Assay Details
Target Gene Details
Entrez Gene ID: | 58493 |
Gene Name: | INTS3 and NABP interacting protein |
Gene Aliases: |
C9orf80, HSPC043, MISE, SOSSC, SSBIP1, hSSBIP1 |
Location: |
Chr.9:112686506-112718107 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INIP | NM_021218.1 | NP_067041.1 | ||
| AF161409.1 | AAF28969.1 | |||
| AF161411.2 | AAF28971.2 | |||
| AK000637.1 | ||||
| AK310007.1 | ||||
| BC014881.1 | AAH14881.1 | |||
| BC065210.1 | AAH65210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv442163 | Chr.9:112620975 - 112828298 on Build GRCh38 | Gain |
|
| nsv615198 | Chr.9:112634926 - 112826971 on Build GRCh38 | Gain |
|
| nsv471317 | Chr.9:112612873 - 112795966 on Build GRCh38 | Gain |
|
| esv2761275 | Chr.9:112614181 - 112828682 on Build GRCh38 | Gain |
|
| esv2759710 | Chr.9:112478174 - 113240671 on Build GRCh38 | Gain |
|
| dgv7701n100 | Chr.9:112604798 - 112828670 on Build GRCh38 | Gain |
|
| esv3621517 | Chr.9:112616823 - 112830602 on Build GRCh38 | Gain |
|
| dgv262e55 | Chr.9:112598225 - 112819173 on Build GRCh38 | Gain |
|
| nsv515121 | Chr.9:112620947 - 112823547 on Build GRCh38 | Gain |
|
| nsv515899 | Chr.9:112596769 - 112831590 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76378582] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map