Assay Details
Target Gene Details
Entrez Gene ID: | 55844 |
Gene Name: | protein phosphatase 2 regulatory subunit Bdelta |
Gene Aliases: |
MDS026 |
Location: |
Chr.10:131900644-131971535 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R2D | XM_006717914.3 | XP_006717977.1 | ||
| XM_017016407.1 | XP_016871896.1 | |||
| AF220046.1 | AAF67639.1 | |||
| AK000402.1 | ||||
| AK293065.1 | ||||
| BC047379.1 | AAH47379.1 | |||
| BC058076.1 | AAH58076.1 | |||
| BC072402.1 | AAH72402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624956 | Chr.10:131249138 - 132223150 on Build GRCh38 | Gain |
|
| nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
|
| nsv825650 | Chr.10:131914064 - 132056715 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map