Assay Details
Target Gene Details
Entrez Gene ID: | 10771 |
Gene Name: | zinc finger MYND-type containing 11 |
Gene Aliases: |
BRAM1, BS69, MRD30 |
Location: |
Chr.10:134340-254637 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMYND11 | NM_001202465.1 | NP_001189394.1 | ||
| NM_001202466.1 | NP_001189395.1 | |||
| NM_006624.5 | NP_006615.2 | |||
| NM_212479.3 | NP_997644.2 | |||
| XM_005252359.4 | XP_005252416.1 | |||
| XM_005252361.3 | XP_005252418.1 | |||
| XM_005252362.2 | XP_005252419.1 | |||
| XM_006717376.2 | XP_006717439.1 | |||
| XM_017015588.1 | XP_016871077.1 | |||
| XM_017015589.1 | XP_016871078.1 | |||
| XM_017015591.1 | XP_016871080.1 | |||
| XM_017015592.1 | XP_016871081.1 | |||
| AK294469.1 | ||||
| AK294781.1 | ||||
| AK312570.1 | ||||
| BC034784.1 | AAH34784.1 | |||
| BP873033.1 | ||||
| DB236591.1 | ||||
| DC376390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622070 | Chr.10:118426 - 174375 on Build GRCh38 | Gain |
|
| dgv117n67 | Chr.10:70307 - 148537 on Build GRCh38 | Gain |
|
| nsv8594 | Chr.10:134401 - 137561 on Build GRCh38 | Gain+Loss |
|
| dgv648n100 | Chr.10:26861 - 157806 on Build GRCh38 | Gain |
|
| nsv1037507 | Chr.10:97603 - 199464 on Build GRCh38 | Gain |
|
| dgv650n100 | Chr.10:83918 - 270899 on Build GRCh38 | Gain |
|
| nsv1069434 | Chr.10:134759 - 135960 on Build GRCh38 | Deletion |
|
| esv2759723 | Chr.10:14061 - 178465 on Build GRCh38 | Gain |
|
| dgv328n106 | Chr.10:134760 - 136060 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map