Assay Details
Target Gene Details
Entrez Gene ID: | 196051 |
Gene Name: | phospholipid phosphatase 4 |
Gene Aliases: |
DPPL2, PPAPDC1, PPAPDC1A |
Location: |
Chr.10:120456954-120600123 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP4 | NM_001030059.2 | NP_001025230.1 | ||
| NM_001318166.1 | NP_001305095.1 | |||
| NM_001318167.1 | NP_001305096.1 | |||
| NM_001318168.1 | NP_001305097.1 | |||
| NM_001318169.1 | NP_001305098.1 | |||
| NR_134516.1 | ||||
| XM_005269592.2 | XP_005269649.1 | |||
| XM_006717686.3 | XP_006717749.1 | |||
| XM_011539444.2 | XP_011537746.1 | |||
| XM_011539445.2 | XP_011537747.1 | |||
| XM_017015823.1 | XP_016871312.1 | |||
| XM_017015824.1 | XP_016871313.1 | |||
| AK296273.1 | ||||
| AK310178.1 | ||||
| BC101267.2 | ||||
| BC101268.2 | ||||
| BC101269.1 | AAI01270.1 | |||
| BC122535.1 | ||||
| BC132787.1 | ||||
| BI550027.1 | ||||
| DB473853.2 | ||||
| DB495310.2 | ||||
| DR000933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv511435 | Chr.10:120466466 - 120471733 on Build GRCh38 | Loss |
|
| dgv1403n54 | Chr.10:120466598 - 120469022 on Build GRCh38 | Gain |
|
| nsv1121823 | Chr.10:120466488 - 120469388 on Build GRCh38 | Deletion |
|
| esv2546781 | Chr.10:120466570 - 120470023 on Build GRCh38 | Deletion |
|
| dgv1402n54 | Chr.10:120457959 - 120469022 on Build GRCh38 | Gain |
|
| nsv436106 | Chr.10:120464890 - 120471999 on Build GRCh38 | Deletion |
|
| esv995047 | Chr.10:120465203 - 120470079 on Build GRCh38 | Deletion |
|
| nsv508610 | Chr.10:120391828 - 120472088 on Build GRCh38 | Deletion |
|
| nsv7572 | Chr.10:120447233 - 120474549 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map