Assay Details
Target Gene Details
Entrez Gene ID: | 340895 |
Gene Name: | MAM and LDL receptor class A domain containing 1 |
Gene Aliases: |
C10orf112, DIET1, bA265G8.2 |
Location: |
Chr.10:19046931-19734478 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MALRD1 | NM_001142308.2 | NP_001135780.2 | ||
| XM_011519453.2 | XP_011517755.1 | |||
| XM_011519454.1 | XP_011517756.1 | |||
| XM_011519455.2 | XP_011517757.1 | |||
| XM_017016182.1 | XP_016871671.1 | |||
| XM_017016183.1 | XP_016871672.1 | |||
| KC843478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv550126 | Chr.10:18981139 - 19283800 on Build GRCh38 | Gain |
|
| esv3891769 | Chr.10:19096319 - 19155253 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map