Assay Details
Target Gene Details
Entrez Gene ID: | 119385 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 11 |
Gene Aliases: |
- |
Location: |
Chr.10:86970741-87010203 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGAP11 | NM_133447.1 | 12 | 2012 | NP_597704.1 |
| AB075855.1 | 7 | 3416 | ||
| BC140762.1 | 8 | 1258 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428236 | Chr.10:86866859 - 87550005 on Build GRCh38 | Loss |
|
| esv27213 | Chr.10:86980917 - 87024746 on Build GRCh38 | Loss |
|
| esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss |
|
| nsv8712 | Chr.10:86997784 - 87027284 on Build GRCh38 | Loss |
|
| nsv831939 | Chr.10:86984277 - 87154254 on Build GRCh38 | Gain |
|
| nsv948111 | Chr.10:86985611 - 87028511 on Build GRCh38 | Duplication |
|
| nsv975708 | Chr.10:87007642 - 87009775 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map