Assay Details
Target Gene Details
Entrez Gene ID: | 387707 |
Gene Name: | coiled-coil and C2 domain containing 2B |
Gene Aliases: |
C10orf130, bA248J23.4 |
Location: |
Chr.10:95974029-96035167 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CC2D2B | XM_011539789.2 | XP_011538091.1 | ||
| XM_011539790.2 | XP_011538092.1 | |||
| XM_011539791.2 | XP_011538093.1 |
Target Gene Details
Entrez Gene ID: | 728558 |
Gene Name: | ENTPD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:95753206-96090235 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD1-AS1 | NR_038444.1 | |||
| AK091396.1 | ||||
| AK096469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624289 | Chr.10:95953322 - 96028771 on Build GRCh38 | Loss |
|
| esv3624287 | Chr.10:95946719 - 95999705 on Build GRCh38 | Loss |
|

Genomic Map