Assay Details
Target Gene Details
Entrez Gene ID: | 120400 |
Gene Name: | neurexophilin and PC-esterase domain family member 1 |
Gene Aliases: |
FAM55A |
Location: |
Chr.11:114518946-114560244 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NXPE1 | NM_152315.3 | NP_689528.2 | ||
| XM_011542594.2 | XP_011540896.1 | |||
| XM_011542595.2 | XP_011540897.1 | |||
| XM_011542596.2 | 1 | 316 | XP_011540898.1 | |
| XM_011542597.2 | 1 | 316 | XP_011540899.1 | |
| XM_011542598.2 | XP_011540900.1 | |||
| XM_011542599.2 | 1 | 316 | XP_011540901.1 | |
| BC029049.1 | AAH29049.1 | |||
| CR747187.1 |
Target Gene Details
Entrez Gene ID: | 120406 |
Gene Name: | neurexophilin and PC-esterase domain family member 2 |
Gene Aliases: |
FAM55B |
Location: |
Chr.11:114464276-114813702 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NXPE2 | XM_017017207.1 | XP_016872696.1 | ||
| XM_017017208.1 | XP_016872697.1 | |||
| XM_017017209.1 | XP_016872698.1 | |||
| XM_017017210.1 | XP_016872699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv512241 | Chr.11:114552961 - 114561276 on Build GRCh38 | Loss |
|
| esv2759865 | Chr.11:114501563 - 114648403 on Build GRCh38 | Gain |
|
| nsv1053425 | Chr.11:114539109 - 114737646 on Build GRCh38 | Loss |
|
| dgv725n106 | Chr.11:114548878 - 114554178 on Build GRCh38 | Duplication |
|
| esv7013 | Chr.11:114553937 - 114561433 on Build GRCh38 | Loss |
|
| nsv952736 | Chr.11:114548879 - 114554178 on Build GRCh38 | Duplication |
|
| nsv975980 | Chr.11:114551091 - 114562531 on Build GRCh38 | Duplication |
|
| nsv832274 | Chr.11:114535182 - 114685944 on Build GRCh38 | Gain |
|
| esv2436910 | Chr.11:114553128 - 114561644 on Build GRCh38 | Deletion |
|
| esv2302049 | Chr.11:114553995 - 114561371 on Build GRCh38 | Deletion |
|

Genomic Map