Assay Details
Target Gene Details
Entrez Gene ID: | 114971 |
Gene Name: | protein tyrosine phosphatase, mitochondrial 1 |
Gene Aliases: |
DUSP23, MOSP, PLIP, PNAS-129 |
Location: |
Chr.11:47565430-47573461 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPMT1 | NM_001143984.1 | NP_001137456.1 | ||
| NM_175732.2 | NP_783859.1 | |||
| AF277187.1 | AAK07545.1 | |||
| AK294640.1 | ||||
| AY333987.1 | AAP94732.1 | |||
| BC014048.1 | AAH14048.1 | |||
| BC020242.1 | AAH20242.1 | |||
| DA476984.1 | ||||
| DR004030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832142 | Chr.11:47496403 - 47659758 on Build GRCh38 | Loss |
|
| nsv825872 | Chr.11:47543873 - 47638778 on Build GRCh38 | Gain |
|
| nsv314 | Chr.11:47540012 - 47584704 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73461796] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map