Assay Details
Target Gene Details
Entrez Gene ID: | 9271 |
Gene Name: | piwi like RNA-mediated gene silencing 1 |
Gene Aliases: |
CT80.1, HIWI, MIWI, PIWI |
Location: |
Chr.12:130337887-130412708 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIWIL1 | XM_017020229.1 | XP_016875718.1 |
Target Gene Details
Entrez Gene ID: | 23504 |
Gene Name: | RIMS binding protein 2 |
Gene Aliases: |
PPP1R133, RBP2, RIM-BP2 |
Location: |
Chr.12:130396133-130716361 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIMBP2 | NM_015347.4 | NP_056162.4 | ||
| XM_011538102.1 | XP_011536404.1 | |||
| XM_011538103.2 | XP_011536405.1 | |||
| XM_011538104.2 | XP_011536406.1 | |||
| XM_011538105.2 | XP_011536407.1 | |||
| XM_011538106.2 | XP_011536408.1 | |||
| XM_017019105.1 | XP_016874594.1 | |||
| XM_017019106.1 | XP_016874595.1 | |||
| XM_017019107.1 | XP_016874596.1 | |||
| XM_017019108.1 | XP_016874597.1 | |||
| XM_017019109.1 | XP_016874598.1 | |||
| XM_017019110.1 | XP_016874599.1 | |||
| XM_017019111.1 | XP_016874600.1 | |||
| AB002316.1 | ||||
| AY699979.1 | ||||
| BX641152.1 | CAE46066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560744 | Chr.12:130395238 - 130790045 on Build GRCh38 | Gain |
|
| nsv818938 | Chr.12:130382306 - 130426671 on Build GRCh38 | Gain |
|
| esv3582027 | Chr.12:130227822 - 130722407 on Build GRCh38 | Gain |
|
| esv3631200 | Chr.12:130375734 - 130607674 on Build GRCh38 | Gain |
|
| esv34208 | Chr.12:129528244 - 130603882 on Build GRCh38 | Gain |
|
| dgv2977n54 | Chr.12:130222462 - 130829669 on Build GRCh38 | Gain |
|
| nsv560735 | Chr.12:129710094 - 130478603 on Build GRCh38 | Gain |
|
| esv2759921 | Chr.12:129461919 - 130631031 on Build GRCh38 | Gain+Loss |
|
| nsv469863 | Chr.12:130350018 - 130554797 on Build GRCh38 | Gain |
|
| nsv1037230 | Chr.12:130378621 - 130425847 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map