Assay Details
Target Gene Details
Entrez Gene ID: | 1272 |
Gene Name: | contactin 1 |
Gene Aliases: |
F3, GP135, MYPCN |
Location: |
Chr.12:40692439-41072412 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTN1 | NM_001256063.1 | NP_001242992.1 | ||
| NM_001843.3 | NP_001834.2 | |||
| XM_017018821.1 | XP_016874310.1 | |||
| AK289698.1 | ||||
| Z21488.1 | CAA79696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832379 | Chr.12:40660638 - 40838947 on Build GRCh38 | Gain |
|
| nsv1048258 | Chr.12:39970608 - 41209299 on Build GRCh38 | Gain |
|
| nsv832378 | Chr.12:40549309 - 40738503 on Build GRCh38 | Gain |
|
| esv2751098 | Chr.12:40688597 - 40786433 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map