Assay Details
Target Gene Details
Entrez Gene ID: | 496 |
Gene Name: | ATPase H+/K+ transporting beta subunit |
Gene Aliases: |
ATP6B |
Location: |
Chr.13:113648804-113658198 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP4B | NM_000705.3 | NP_000696.1 | ||
| BC029059.1 | AAH29059.1 | |||
| BM923761.1 | ||||
| M75110.1 | AAA35987.1 |
Target Gene Details
Entrez Gene ID: | 6011 |
Gene Name: | G protein-coupled receptor kinase 1 |
Gene Aliases: |
GPRK1, RHOK, RK |
Location: |
Chr.13:113645771-113735664 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK1 | XM_017020684.1 | XP_016876173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832726 | Chr.13:113559093 - 113663088 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map