Assay Details
Target Gene Details
Entrez Gene ID: | 23077 |
Gene Name: | MYC binding protein 2, E3 ubiquitin protein ligase |
Gene Aliases: |
Myc-bp2, PAM, Phr |
Location: |
Chr.13:77044657-77327066 on Build GRCh38 |
Assay Gene Location: | Within Intron 71 |
Target Gene Details
Entrez Gene ID: | 100874212 |
Gene Name: | MYCBP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:77075514-77087776 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCBP2-AS1 | NR_046716.1 | |||
| BG208820.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832653 | Chr.13:76948899 - 77115221 on Build GRCh38 | Loss |
|
| esv2759951 | Chr.13:77050897 - 77194329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map