Assay Details
Target Gene Details
Entrez Gene ID: | 101929140 |
Gene Name: | uncharacterized LOC101929140 |
Gene Aliases: |
- |
Location: |
Chr.13:41132928-41236686 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929140 | NR_120423.1 | |||
| AK056182.1 | ||||
| DA634395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832587 | Chr.13:41030795 - 41224672 on Build GRCh38 | Gain |
|
| esv3631867 | Chr.13:41144449 - 41179962 on Build GRCh38 | Gain |
|
| esv2747299 | Chr.13:41131549 - 41193448 on Build GRCh38 | Deletion |
|
| nsv983580 | Chr.13:41149809 - 41173713 on Build GRCh38 | Duplication |
|
| nsv523358 | Chr.13:41144691 - 41186811 on Build GRCh38 | Loss |
|
| nsv528180 | Chr.13:41141146 - 41186811 on Build GRCh38 | Gain |
|

Genomic Map