Assay Details
Target Gene Details
Entrez Gene ID: | 113622 |
Gene Name: | ADP-ribosylhydrolase like 1 |
Gene Aliases: |
ARH2 |
Location: |
Chr.13:113421940-113453524 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADPRHL1 | NM_138430.4 | NP_612439.2 | ||
| NM_199162.2 | NP_954631.1 | |||
| AJ313429.1 | CAC86114.1 | |||
| BC009755.2 | AAH09755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048814 | Chr.13:113081728 - 113611496 on Build GRCh38 | Gain |
|
| dgv1744n100 | Chr.13:113384831 - 113442576 on Build GRCh38 | Gain |
|
| nsv563321 | Chr.13:113369086 - 113452383 on Build GRCh38 | Loss |
|
| nsv9116 | Chr.13:113434198 - 113446711 on Build GRCh38 | Gain |
|
| esv2759970 | Chr.13:113384114 - 113556749 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113376456] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map