Assay Details
Target Gene Details
Entrez Gene ID: | 91833 |
Gene Name: | WD repeat domain 20 |
Gene Aliases: |
DMR |
Location: |
Chr.14:102139518-102224686 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565947 | Chr.14:102001510 - 102204240 on Build GRCh38 | Gain |
|
| dgv25n68 | Chr.14:102000993 - 102195522 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map