Assay Details
Target Gene Details
Entrez Gene ID: | 11154 |
Gene Name: | adaptor related protein complex 4 sigma 1 subunit |
Gene Aliases: |
AP47B, CLA20, CLAPS4, CPSQ6, SPG52 |
Location: |
Chr.14:31025106-31096450 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4S1 | NM_001128126.2 | NP_001121598.1 | ||
| NM_001254726.1 | NP_001241655.1 | |||
| NM_001254727.1 | NP_001241656.1 | |||
| NM_001254728.1 | NP_001241657.1 | |||
| NM_001254729.1 | NP_001241658.1 | |||
| NM_007077.4 | NP_009008.2 | |||
| XM_005267293.4 | XP_005267350.1 | |||
| XM_011536371.2 | XP_011534673.1 | |||
| XM_011536372.2 | XP_011534674.1 | |||
| AB030654.1 | BAA82970.1 | |||
| AF155159.1 | AAD43329.1 | |||
| AK304115.1 | ||||
| AK316517.1 | ||||
| BC001259.1 | AAH01259.1 | |||
| BF671561.1 | ||||
| BG702035.1 | ||||
| BX247969.1 | CAD62307.1 | |||
| BX388185.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564169 | Chr.14:30349881 - 31072288 on Build GRCh38 | Gain |
|
| nsv1233 | Chr.14:31023799 - 31035069 on Build GRCh38 | Insertion |
|
| nsv1049063 | Chr.14:31004315 - 31153535 on Build GRCh38 | Gain |
|
| esv3892495 | Chr.14:30985830 - 31116083 on Build GRCh38 | Gain |
|
| esv33189 | Chr.14:30331017 - 31442539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map