Assay Details
Target Gene Details
Entrez Gene ID: | 90673 |
Gene Name: | protein phosphatase 1 regulatory subunit 3E |
Gene Aliases: |
- |
Location: |
Chr.14:23295921-23302848 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R3E | NM_001276318.1 | 5 | 1686 | NP_001263247.1 |
| AK024489.1 | 5 | 1686 | ||
| AK091943.1 | ||||
| AK296016.1 | 3 | 454 | ||
| BX161476.1 | 3 | 1149 | CAD61931.1 | |
| BX248758.1 | 3 | 1163 | CAD66565.1 |
More Information
Set Membership: |
|

Genomic Map