Assay Details
Target Gene Details
Entrez Gene ID: | 400410 |
Gene Name: | suppressor of tumorigenicity 20 |
Gene Aliases: |
HCCS-1 |
Location: |
Chr.15:79898840-79923754 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST20 | NM_001100879.1 | NP_001094349.1 | ||
| NM_001100880.2 | NP_001094350.1 | |||
| NM_001199757.1 | NP_001186686.1 | |||
| NR_037652.1 | ||||
| NR_037653.1 | ||||
| AA282293.1 | ||||
| AF249277.1 | AAG17280.1 | |||
| AL109714.1 | ||||
| BM559302.1 | ||||
| BQ680929.1 |
Target Gene Details
Entrez Gene ID: | 100528021 |
Gene Name: | ST20-MTHFS readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:79843547-79923106 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST20-MTHFS | NM_001199760.1 | NP_001186689.1 | ||
| BG705061.1 | ||||
| CK002935.1 | ||||
| DB495490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833066 | Chr.15:79878078 - 80033241 on Build GRCh38 | Loss |
|
| esv3637009 | Chr.15:79898247 - 79960449 on Build GRCh38 | Loss |
|
| nsv833065 | Chr.15:79760661 - 79982480 on Build GRCh38 | Loss |
|
| esv2660680 | Chr.15:79898180 - 79960294 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map