Assay Details
Target Gene Details
Entrez Gene ID: | 25820 |
Gene Name: | ariadne RBR E3 ubiquitin protein ligase 1 |
Gene Aliases: |
ARI, HARI, HHARI, UBCH7BP |
Location: |
Chr.15:72474282-72586563 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARIH1 | NM_005744.3 | NP_005735.2 | ||
| XM_011521438.2 | XP_011519740.1 | |||
| XM_011521439.2 | XP_011519741.1 | |||
| AB209432.1 | BAD92669.1 | |||
| AF072832.1 | AAD28088.1 | |||
| AJ130976.1 | CAA10274.1 | |||
| AJ243190.1 | CAB45870.1 | |||
| AK312715.1 | ||||
| BC051877.1 | AAH51877.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469704 | Chr.15:72377033 - 72561586 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map