Assay Details
Target Gene Details
Entrez Gene ID: | 145853 |
Gene Name: | chromosome 15 open reading frame 61 |
Gene Aliases: |
- |
Location: |
Chr.15:67521184-67527303 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C15orf61 | NM_001143936.1 | 2 | 905 | NP_001137408.1 |
| AK294012.1 | 2 | 840 | ||
| BM664696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569799 | Chr.15:67511204 - 67585107 on Build GRCh38 | Loss |
|

Genomic Map