Assay Details
Target Gene Details
Entrez Gene ID: | 2232 |
Gene Name: | ferredoxin reductase |
Gene Aliases: |
ADXR |
Location: |
Chr.17:74862497-74873031 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FDXR | NM_001258012.3 | NP_001244941.2 | ||
| NM_001258013.3 | NP_001244942.2 | |||
| NM_001258014.3 | NP_001244943.2 | |||
| NM_001258015.2 | NP_001244944.1 | |||
| NM_004110.5 | NP_004101.3 | |||
| NM_024417.4 | NP_077728.3 | |||
| NR_047576.2 | ||||
| XM_006721772.1 | XP_006721835.1 | |||
| XM_011524527.1 | XP_011522829.1 | |||
| XM_011524528.1 | XP_011522830.1 | |||
| XM_011524529.1 | XP_011522831.1 | |||
| AK094120.1 | ||||
| AK098648.1 | ||||
| AK223530.1 | BAD97250.1 | |||
| AK293208.1 | ||||
| AK293210.1 | ||||
| AK295307.1 | ||||
| AK298908.1 | ||||
| AK301779.1 | ||||
| BC002960.1 | AAH02960.1 | |||
| BC063493.1 | AAH63493.1 | |||
| BT006684.1 | AAP35330.1 | |||
| DA267443.1 | ||||
| J03826.1 | AAB59497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map