Assay Details
Target Gene Details
Entrez Gene ID: | 84667 |
Gene Name: | hes family bHLH transcription factor 7 |
Gene Aliases: |
SCDO4, bHLHb37 |
Location: |
Chr.17:8120590-8126032 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HES7 | XM_011524041.2 | XP_011522343.1 | ||
| XM_017025232.1 | XP_016880721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518983 | Chr.17:8124275 - 8147661 on Build GRCh38 | Loss |
|
| nsv833352 | Chr.17:8029719 - 8187575 on Build GRCh38 | Loss |
|
| nsv457661 | Chr.17:8106586 - 8147661 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map