Assay Details
Target Gene Details
Entrez Gene ID: | 201140 |
Gene Name: | dehydrogenase/reductase 7C |
Gene Aliases: |
SDR32C2 |
Location: |
Chr.17:9771434-9791297 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS7C | NM_001105571.2 | NP_001099041.1 | ||
| NM_001220493.1 | NP_001207422.1 | |||
| BC147024.1 | ||||
| BC147025.1 | ||||
| BC171909.1 | ||||
| DN831946.1 | ||||
| DN831947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833358 | Chr.17:9643743 - 9828246 on Build GRCh38 | Gain |
|
| nsv833360 | Chr.17:9734634 - 9900986 on Build GRCh38 | Loss |
|
| esv3639923 | Chr.17:9778120 - 9786051 on Build GRCh38 | Loss |
|
| nsv510697 | Chr.17:9729641 - 9812090 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map