Assay Details
Target Gene Details
Entrez Gene ID: | 7531 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon |
Gene Aliases: |
14-3-3E, HEL2, KCIP-1, MDCR, MDS |
Location: |
Chr.17:1344490-1400375 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YWHAE | NM_006761.4 | NP_006752.1 | ||
| NR_024058.1 | ||||
| XM_005256784.3 | XP_005256841.1 | |||
| XM_017025005.1 | XP_016880494.1 | |||
| AK128785.1 | ||||
| AK295260.1 | ||||
| AK295997.1 | ||||
| AK296050.1 | ||||
| AK316185.1 | ||||
| AK316201.1 | ||||
| AY883089.1 | AAX68683.1 | |||
| BC000179.2 | AAH00179.1 | |||
| BC001440.2 | AAH01440.1 | |||
| BT007161.1 | AAP35825.1 | |||
| EU794581.1 | ||||
| U20972.1 | AAC50175.1 | |||
| U28936.1 | AAA75301.1 | |||
| U43399.1 | AAC50625.1 | |||
| U43430.1 | AAD00026.1 | |||
| U54778.1 | AAC50710.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3081n100 | Chr.17:1095450 - 1360904 on Build GRCh38 | Gain |
|
| nsv528360 | Chr.17:1330341 - 1365389 on Build GRCh38 | Gain |
|
| nsv574209 | Chr.17:1341698 - 1405534 on Build GRCh38 | Loss |
|
| nsv827854 | Chr.17:1331993 - 1357097 on Build GRCh38 | Gain |
|
| nsv1057782 | Chr.17:1351585 - 1512616 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map