Assay Details
Target Gene Details
Entrez Gene ID: | 5148 |
Gene Name: | phosphodiesterase 6G |
Gene Aliases: |
PDEG, RP57 |
Location: |
Chr.17:81650459-81663418 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE6G | XM_017024734.1 | XP_016880223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828128 | Chr.17:80659041 - 81736487 on Build GRCh38 | Loss |
|
| nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain |
|
| nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss |
|
| nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs68177983] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map