Assay Details
Target Gene Details
Entrez Gene ID: | 149775 |
Gene Name: | GNAS antisense RNA 1 |
Gene Aliases: |
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG |
Location: |
Chr.20:58818918-58850903 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAS-AS1 | NR_002785.2 | |||
| AJ251759.1 | ||||
| BF589977.1 | ||||
| DB067618.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76473307] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map