Assay Details
Target Gene Details
Entrez Gene ID: | 90199 |
Gene Name: | WAP four-disulfide core domain 8 |
Gene Aliases: |
C20orf170, HEL-S-292, WAP8, dJ461P17.1 |
Location: |
Chr.20:45551152-45579326 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC8 | NM_130896.2 | NP_570966.2 | ||
| NM_181510.2 | NP_852611.2 | |||
| XM_017028119.1 | XP_016883608.1 | |||
| AF492015.1 | AAN70997.1 | |||
| AF492016.1 | AAN70998.1 | |||
| GQ472209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2661076 | Chr.20:45575422 - 45578660 on Build GRCh38 | Deletion |
|
| esv25639 | Chr.20:45575598 - 45578663 on Build GRCh38 | Loss |
|
| esv3557118 | Chr.20:45575334 - 45578700 on Build GRCh38 | Deletion |
|
| dgv7587n54 | Chr.20:45575651 - 45577280 on Build GRCh38 | Loss |
|
| nsv586080 | Chr.20:45573240 - 45578612 on Build GRCh38 | Loss |
|
| esv2722448 | Chr.20:45575374 - 45578646 on Build GRCh38 | Deletion |
|
| esv32588 | Chr.20:45575744 - 45579160 on Build GRCh38 | Loss |
|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
| esv3645935 | Chr.20:45575405 - 45578675 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map