Assay Details
Target Gene Details
Entrez Gene ID: | 10738 |
Gene Name: | ret finger protein like 3 |
Gene Aliases: |
- |
Location: |
Chr.22:32354885-32361161 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFPL3 | XM_017028534.1 | XP_016884023.1 | ||
| AK097738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575476 | Chr.22:32073030 - 32383792 on Build GRCh38 | Gain |
|
| esv3893474 | Chr.22:32054375 - 32387314 on Build GRCh38 | Gain |
|
| nsv588895 | Chr.22:32178287 - 32470947 on Build GRCh38 | Gain |
|
| nsv964624 | Chr.22:32344331 - 32355919 on Build GRCh38 | Duplication |
|
| esv2422247 | Chr.22:32090461 - 32392271 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115825524,rs78134052] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map