Assay Details
Target Gene Details
Entrez Gene ID: | 10739 |
Gene Name: | ret finger protein like 2 |
Gene Aliases: |
RNF79 |
Location: |
Chr.22:32190435-32204844 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFPL2 | NM_001098527.2 | NP_001091997.2 | ||
| NM_001159545.1 | NP_001153017.1 | |||
| NM_001159546.1 | NP_001153018.1 | |||
| XM_011529826.1 | XP_011528128.1 | |||
| XM_011529827.2 | XP_011528129.1 | |||
| XM_011529828.1 | XP_011528130.1 | |||
| XM_011529830.2 | XP_011528132.1 | |||
| XM_011529832.1 | XP_011528134.1 | |||
| XM_017028535.1 | XP_016884024.1 | |||
| XM_017028536.1 | XP_016884025.1 | |||
| AK055999.1 | ||||
| BC051910.1 | AAH51910.1 | |||
| CR456563.1 | CAG30449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575476 | Chr.22:32073030 - 32383792 on Build GRCh38 | Gain |
|
| esv3893474 | Chr.22:32054375 - 32387314 on Build GRCh38 | Gain |
|
| nsv588895 | Chr.22:32178287 - 32470947 on Build GRCh38 | Gain |
|
| nsv953071 | Chr.22:32198214 - 32213613 on Build GRCh38 | Deletion |
|
| nsv834178 | Chr.22:32094462 - 32281534 on Build GRCh38 | Loss |
|
| esv2422247 | Chr.22:32090461 - 32392271 on Build GRCh38 | Duplication |
|
| nsv3597 | Chr.22:32192437 - 32236737 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map