Assay Details
Target Gene Details
Entrez Gene ID: | 286464 |
Gene Name: | cilia and flagella associated protein 47 |
Gene Aliases: |
CHDC2, CXorf22, CXorf30, CXorf59 |
Location: |
Chr.X:35919734-36385319 on Build GRCh38 |
Assay Gene Location: | Within Intron 63 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP47 | NM_001304548.1 | NP_001291477.1 | ||
| AI822140.1 | ||||
| BG190566.1 | ||||
| BG197183.1 |
Target Gene Details
Entrez Gene ID: | 101928627 |
Gene Name: | uncharacterized LOC101928627 |
Gene Aliases: |
- |
Location: |
Chr.X:36365626-36440292 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928627 | NR_110412.1 | |||
| AK092146.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470349 | Chr.X:35988125 - 36420457 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map