Assay Details
Target Gene Details
Entrez Gene ID: | 389860 |
Gene Name: | PAGE family member 2B |
Gene Aliases: |
CT16.5, GAGEE3, PAGE-2B |
Location: |
Chr.X:55027927-55078903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAGE2B | XM_011530785.2 | XP_011529087.1 | ||
| XM_011530786.2 | XP_011529088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv1132768 | Chr.X:55034767 - 55037667 on Build GRCh38 | Duplication |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|

Genomic Map