Assay Details
Target Gene Details
Entrez Gene ID: | 347442 |
Gene Name: | DDB1 and CUL4 associated factor 8 like 2 |
Gene Aliases: |
WDR42C |
Location: |
Chr.X:27746809-27748821 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF8L2 | NM_001136533.1 | 1 | 770 | NP_001130005.1 |
| BC157859.1 | 1 | 770 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508756 | Chr.X:27744422 - 27779117 on Build GRCh38 | Insertion |
|
| nsv521729 | Chr.X:27692376 - 27821455 on Build GRCh38 | Gain |
|
| nsv526948 | Chr.X:27464175 - 27901289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map