Assay Details
Target Gene Details
Entrez Gene ID: | 824 |
Gene Name: | calpain 2 |
Gene Aliases: |
CANP2, CANPL2, CANPml, mCANP |
Location: |
Chr.1:223701593-223776018 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPN2 | NM_001146068.1 | NP_001139540.1 | ||
| AK297800.1 | ||||
| AK316211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34082 | Chr.1:223663316 - 223956807 on Build GRCh38 | Loss |
|
| nsv832681 | Chr.1:223524706 - 223814989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map