Assay Details
Target Gene Details
Entrez Gene ID: | 11124 |
Gene Name: | Fas associated factor 1 |
Gene Aliases: |
CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1 |
Location: |
Chr.1:50441263-50960264 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAF1 | NM_007051.2 | NP_008982.1 | ||
| XM_017000136.1 | XP_016855625.1 | |||
| AF094700.1 | AAD51876.1 | |||
| AF106798.1 | AAD51886.1 | |||
| AF132938.1 | AAD27713.1 | |||
| AF136173.1 | AAP97263.1 | |||
| AJ271408.1 | CAB67705.1 | |||
| AK094843.1 | ||||
| AK292840.1 | ||||
| AK293659.1 | ||||
| AL133631.1 | CAB63755.1 | |||
| BC004970.1 | AAH04970.1 | |||
| BC067100.1 | AAH67100.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115069774] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map