Assay Details
Target Gene Details
Entrez Gene ID: | 128153 |
Gene Name: | spermatogenesis associated 17 |
Gene Aliases: |
IQCH, MSRG-11, MSRG11 |
Location: |
Chr.1:217631324-217868151 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA17 | NM_138796.3 | NP_620151.1 | ||
| XM_005273052.1 | XP_005273109.1 | |||
| XM_006711165.3 | XP_006711228.1 | |||
| XM_011509194.2 | XP_011507496.1 | |||
| AK098591.1 | BAC05343.1 | |||
| AK310291.1 | ||||
| AY963797.1 | ||||
| BC014608.2 | AAH14608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762166 | Chr.1:217588526 - 217727313 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map