Assay Details
Target Gene Details
Entrez Gene ID: | 64756 |
Gene Name: | ATP synthase mitochondrial F1 complex assembly factor 1 |
Gene Aliases: |
ATP11, ATP11p |
Location: |
Chr.1:46635039-46668427 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATPAF1 | NM_001042546.2 | NP_001036011.2 | ||
| NM_001243728.1 | NP_001230657.1 | |||
| NM_001256418.1 | NP_001243347.1 | |||
| NM_022745.4 | NP_073582.3 | |||
| AK026004.1 | BAB15316.1 | |||
| AK301874.1 | ||||
| AK302090.1 | ||||
| AK310338.1 | ||||
| AK311271.1 | ||||
| BC008498.1 | AAH08498.1 | |||
| BC018781.1 | AAH18781.1 | |||
| BC071731.1 | AAH71731.1 | |||
| BX438342.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007280 | Chr.1:46622444 - 46663916 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map