Assay Details
Target Gene Details
Entrez Gene ID: | 5293 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta |
Gene Aliases: |
APDS, IMD14, P110DELTA, PI3K, p110D |
Location: |
Chr.1:9629889-9729114 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CD | XM_017001477.1 | XP_016856966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508847 | Chr.1:9612137 - 9678831 on Build GRCh38 | Insertion |
|
| nsv2187 | Chr.1:9607238 - 9633692 on Build GRCh38 | Insertion |
|
| nsv1013224 | Chr.1:9380981 - 9639710 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map