Assay Details
Target Gene Details
Entrez Gene ID: | 978 |
Gene Name: | cytidine deaminase |
Gene Aliases: |
CDD |
Location: |
Chr.1:20588951-20618908 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDA | NM_001785.2 | NP_001776.1 | ||
| AJ000474.1 | CAA04113.1 | |||
| AY634312.2 | ||||
| BC054036.1 | AAH54036.1 | |||
| CD675458.1 | ||||
| L27943.1 | AAA57254.1 | |||
| S52873.1 | AAB24946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
| nsv528156 | Chr.1:20589208 - 20608303 on Build GRCh38 | Loss |
|
| esv998962 | Chr.1:20598896 - 20604674 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map