Assay Details
Target Gene Details
Entrez Gene ID: | 117145 |
Gene Name: | thioesterase superfamily member 4 |
Gene Aliases: |
CTMP |
Location: |
Chr.1:151870866-151909885 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THEM4 | NM_053055.4 | NP_444283.2 | ||
| XM_011509142.2 | XP_011507444.1 | |||
| AJ313515.1 | CAC86384.1 | |||
| AK289494.1 | ||||
| AK314852.1 | ||||
| BC065277.1 | AAH65277.1 | |||
| BM787449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547852 | Chr.1:151870568 - 151982754 on Build GRCh38 | Loss |
|
| nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map