Assay Details
Target Gene Details
Entrez Gene ID: | 25896 |
Gene Name: | integrator complex subunit 7 |
Gene Aliases: |
C1orf73, INT7 |
Location: |
Chr.1:211940328-212035660 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS7 | NM_001199809.1 | 19 | 4284 | NP_001186738.1 |
| NM_001199811.1 | 20 | 4389 | NP_001186740.1 | |
| NM_001199812.1 | 20 | 4371 | NP_001186741.1 | |
| NM_015434.3 | 20 | 4431 | NP_056249.1 | |
| NR_037667.1 | 21 | 4465 | ||
| XM_011509396.2 | 21 | 4424 | XP_011507698.1 | |
| AL133049.2 | 15 | 3600 | CAB61376.1 |
More Information
Set Membership: |
|

Genomic Map