Assay Details
Target Gene Details
Entrez Gene ID: | 8863 |
Gene Name: | period circadian clock 3 |
Gene Aliases: |
FASPS3, GIG13 |
Location: |
Chr.1:7784285-7845181 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
More Information
Set Membership: |
|

Genomic Map