Assay Details
Target Gene Details
Entrez Gene ID: | 192669 |
Gene Name: | argonaute 3, RISC catalytic component |
Gene Aliases: |
EIF2C3 |
Location: |
Chr.1:35925198-36072037 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGO3 | NM_024852.3 | NP_079128.2 | ||
NM_177422.2 | NP_803171.1 | |||
XM_005270575.3 | XP_005270632.1 | |||
XM_011540879.2 | XP_011539181.1 | |||
XM_017000523.1 | XP_016856012.1 | |||
AK022827.1 | BAB14262.1 | |||
AK027796.1 | BAB55374.1 | |||
AK292870.1 | ||||
BC066888.1 | AAH66888.1 | |||
BF979526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv998713 | Chr.1:35956942 - 36008597 on Build GRCh38 | Gain |
![]() |
dgv149n100 | Chr.1:35947159 - 36032074 on Build GRCh38 | Loss |
![]() |
nsv1009992 | Chr.1:35949901 - 36009793 on Build GRCh38 | Gain+Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
