Assay Details
Target Gene Details
Entrez Gene ID: | 9019 |
Gene Name: | myelin protein zero like 1 |
Gene Aliases: |
MPZL1b, PZR, PZR1b, PZRa, PZRb |
Location: |
Chr.1:167721950-167791919 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPZL1 | NM_001146191.1 | NP_001139663.1 | ||
| NM_003953.5 | NP_003944.1 | |||
| NM_024569.4 | NP_078845.3 | |||
| AF087020.1 | AAC72231.1 | |||
| AF092425.1 | AAD55347.1 | |||
| AF095726.1 | AAF00083.1 | |||
| AF095727.1 | AAF00084.1 | |||
| AF181660.1 | AAF00621.1 | |||
| AF239756.1 | AAF63499.1 | |||
| AF478447.1 | AAO14645.1 | |||
| AK075334.1 | ||||
| AK291249.1 | ||||
| AK297112.1 | ||||
| AK307250.1 | ||||
| AK315546.1 | ||||
| AY359019.1 | AAQ89378.1 | |||
| BC007881.2 | AAH07881.1 | |||
| BE616859.1 | ||||
| BF528837.1 | ||||
| BT019542.1 | AAV38349.1 | |||
| CR542160.1 | CAG46957.1 | |||
| DA725074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587852 | Chr.1:167735812 - 167738308 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map