Assay Details
Target Gene Details
Entrez Gene ID: | 9111 |
Gene Name: | N-myc and STAT interactor |
Gene Aliases: |
- |
Location: |
Chr.2:151270465-151289916 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NMI | NM_004688.2 | NP_004679.2 | ||
| XM_005246941.2 | XP_005246998.1 | |||
| XM_017005247.1 | XP_016860736.1 | |||
| AK124323.1 | ||||
| AK291548.1 | ||||
| AK298754.1 | ||||
| BC001268.1 | AAH01268.1 | |||
| CB151555.1 | ||||
| U32849.1 | AAC12949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34659 | Chr.2:150858366 - 152860232 on Build GRCh38 | Loss |
|
| nsv1012891 | Chr.2:151163295 - 151450383 on Build GRCh38 | Gain |
|
| nsv834417 | Chr.2:151123959 - 151298436 on Build GRCh38 | Gain |
|
| dgv4094n100 | Chr.2:151258123 - 151619882 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map