Assay Details
Target Gene Details
Entrez Gene ID: | 4084 |
Gene Name: | MAX dimerization protein 1 |
Gene Aliases: |
BHLHC58, MAD, MAD1 |
Location: |
Chr.2:69915041-69942945 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MXD1 | NM_001202513.1 | NP_001189442.1 | ||
| NM_001202514.1 | NP_001189443.1 | |||
| NM_002357.3 | NP_002348.1 | |||
| AK301088.1 | ||||
| AK312734.1 | ||||
| BC069377.1 | AAH69377.1 | |||
| BC069433.1 | AAH69433.1 | |||
| BC098396.1 | AAH98396.1 | |||
| BC113531.1 | ||||
| BC117260.1 | ||||
| BC143831.1 | ||||
| BC143832.1 | ||||
| CR536495.1 | CAG38734.1 | |||
| CR541692.1 | CAG46493.1 | |||
| DC347823.1 | ||||
| L06895.1 | AAA36194.1 |
More Information
Set Membership: |
|

Genomic Map