Assay Details
Target Gene Details
Entrez Gene ID: | 91526 |
Gene Name: | ankyrin repeat domain 44 |
Gene Aliases: |
PP6-ARS-B |
Location: |
Chr.2:196962617-197310797 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD44 | NM_001195144.1 | NP_001182073.1 | ||
| NM_153697.2 | NP_710181.2 | |||
| XM_005246945.1 | XP_005247002.1 | |||
| XM_005246946.1 | XP_005247003.1 | |||
| XM_005246947.2 | XP_005247004.1 | |||
| XM_005246948.2 | XP_005247005.1 | |||
| XM_006712838.3 | XP_006712901.1 | |||
| XM_011512148.2 | XP_011510450.1 | |||
| XM_017005252.1 | XP_016860741.1 | |||
| AK124039.1 | ||||
| AK301209.1 | ||||
| BC016985.2 | AAH16985.2 | |||
| BC063622.1 | AAH63622.1 |
Target Gene Details
Entrez Gene ID: | 101927547 |
Gene Name: | ANKRD44 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.2:197250858-197302519 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD44-IT1 | NR_046837.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4149n100 | Chr.2:197281159 - 197371942 on Build GRCh38 | Gain |
|
| nsv1002666 | Chr.2:197183292 - 197300397 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map