Assay Details
Target Gene Details
Entrez Gene ID: | 9655 |
Gene Name: | suppressor of cytokine signaling 5 |
Gene Aliases: |
CIS6, CISH6, Cish5, SOCS-5 |
Location: |
Chr.2:46698960-46762788 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS5 | NM_014011.4 | NP_054730.1 | ||
| NM_144949.2 | NP_659198.1 | |||
| AB014571.1 | ||||
| AF073958.1 | AAD40484.1 | |||
| AK290194.1 | ||||
| AK296796.1 | ||||
| AL136896.1 | CAB66830.1 | |||
| BC032862.1 | AAH32862.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013079 | Chr.2:46722198 - 46771278 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map